Order Entry
Belgium
ContactUsLinkComponent
Lovibond® SD10 Water Testing pH Pocket Tester
Lovibond® SD10 Water Testing pH Pocket Tester
Catalog # LOVI720010
Supplier:  Lovibond
undefined
Lovibond® SD10 Water Testing pH Pocket Tester
Catalog # LOVI720010
Supplier:  Lovibond

Specifications

  • Description:
    SD10 pH Pocket tester
  • Model:
    SD10
  • pH range:
    pH: ± 0.01 (±1 digit)
  • Pk:
    1

Specifications

About this item

SD10 pH with its waterproof housing, the pocket tester is ideal for regular checks in industrial plants or swimming pools.

  • Resolution pH: 0.01 ;pH (mV): 1
  • Resolution Temperature: 0,1 °C
  • Calibration Points 1 to 3
  • pH: 0 to 14
  • pH (mV) : 1000 to 1000
  • Temperature: 0 to 50 ° C
  • Dimensions: 40×40×180 mm
  • Weight: 151 g
  • Weight with Packaging: 268 g

Plus, the transparent lid not only provides a secure stand for the Pocket Tester in daily use, but also makes it easier to maintain the electrodes. A three-colour LED lighting system - blue, green and red - and new display icons make the tester easy to use and intuitive to operate. The use of energy saving LEDs and 4× AAA batteries greatly extends the life of the new Pocket Tester range to up to 2000 hours

Application: Boiler Water, cooling water, drinking water, treatment, pool water-control, waste water treatment.

With the included lanyard, each SD Series tester can be worn comfortably around the neck or attached to a pocket, waistband or belt with the clip on the head of the tester, allowing it to be taken anywhere
without tiring the hands.

Calibration indicator: Yes.
Stability indicator: Yes.
Auto-hold function: Yes.
Alarm limits: Yes.
Environmental Conditions: 0 to 60 °C/0 to 80% rel. humidity (non-condensing).

Certifications: Class IP 67, Compliance CE.
Delivery: It include meter in a robust plastic case with hanger, 4 batteries, lanyard, pH 4, 7, 10 buffer tablets (1 strip of 10 tablets each), 3 M KCl storage solution for pH/ORP, instruction manual, warranty declaration, certificate of compliance.