Orders LinkContactUsLinkComponent
Broad range hydrometers
Broad range hydrometers
Broad range hydrometers
Broad range hydrometers
Broad range hydrometers
These broad range hydrometers are calibrated at 20 °C.
Mostly dedicated to applications with citric and tartaric acids.
Product Family Options
  • Range
    Division
    Length
  • 1,000 − 2,000 °Bé
    0,01
    300 mm
    1 * 1 ST
    1 * 1 ST - 21,70 €
    21,70 €
    1 * 1 ST
       21,70 €
  • 0,650 − 1,000 °Bé
    0,005
    300 mm
    1 * 1 ST
    1 * 1 ST - 21,70 €
    21,70 €
    1 * 1 ST
       21,70 €
  • 1,000 − 1,500 °Bé
    0,005
    235 mm
    1 * 1 ST
    1 * 1 ST - 43,20 €
    43,20 €
    1 * 1 ST
       43,20 €