Order Entry
Belgium
ContactUsLinkComponent
Acetic acid glacial
Flammable
Corrosive
Acetic acid glacial
Catalog # VWRS5976.9210
Supplier:  VWR Collection
CAS Number:  
undefined
Flammable
Corrosive
Acetic acid glacial
Catalog # VWRS5976.9210
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    210 L
  • Seq:
    5585

Specifications

About this item

 :
 :60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
 :GHS02
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152