

2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride colorimetric assays, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:146-68-9
:
Formula:C₁₉H₁₃ClIN₅O₂
MDL Number:MFCD00011961
:505.7 g/mol
Melting Pt:227…229 °C
:M-Clarity™ quality level = MQ200
EINECS:205-676-2
:GHS02
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance (Colour):Light Yellow to Yellow
- Appearance (Form):Powder
- Solubility (Colour):Faint Green to Light Green and Faint Green-Yellow to Green-Yellow and Light Yellow to Yellow
- Solubility (Turbidity)
4 mg/ml, H₂O:Clear - Solvent Content (by GC):≤4 %
- Purity (TLC):≥98 %
- Recommended Retest Period:5 years
Specification Test Results
Frequently Bought Together