


Formaldehyde solution 35% (w/w) stabilised, meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
Boiling Pt:93…96 °C (1.013 hPa)
Melting Pt:<–15 °C
Density:1.080…1.090 g/cm³ (20 °C)
Flash Pt:64 °C
Storage Temperature:Ambient
UN:2209
ADR:8,III
:Stabilised with methanol 10%
EINECS:200-001-8
:GHS05
:GHS08
:GHS06
Specification Test Results
- Assay:35.0 - 38.0 %
- Density (D 20/4):effective
- Sulfated ash:Max. 0.01 %
- Acidity:complying
- Methanol (GC):9.0 - 15.0 Vol.%
- Residual solvents:complying
- Appearance of the solution:complying
- Autoignition Temperature:572 °F
- Vapor Pressure:52 mmHg ( 37 °C)
Specification Test Results
Frequently Bought Together