Thymine
Glass bottle for solids, 50 g
Catalog # SERA36460.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
- CAS Number:65-71-4
- Formula:C₅H₆N₂O₂
- MDL Number:MFCD00006026
- Molecular Weight:126.11 g/mol
- Merck Index:12,09539
- Boiling Pt:389 °C (1013 hPa)
- Melting Pt:218 °C
- Density:1.454 g/cm³ (25 °C)
- EINECS:200-616-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



