2-Deoxy-D(-)-ribose
Glass bottle for solids, 5 g
Catalog # SERA18590.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: 2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal
- CAS Number:533-67-5
- Formula:C₅H₁₀O₄
- MDL Number:MFCD00135904
- Molecular Weight:134.13 g/mol
- Merck Index:13,02925
- Boiling Pt:292 °C (1013 hPa)
- Melting Pt:95…97 °C
- Storage Temperature:Ambient
- EINECS:208-573-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



