Order Entry
Belgium
ContactUsLinkComponent
Potassium dichromate 0.02 mol/l (0.12 N) in aqueous solution
Toxic
Health
Potassium dichromate 0.02 mol/l (0.12 N) in aqueous solution
Catalog # ROTHP720.1
Supplier:  Roth Carl
CAS Number:  
Toxic
Health
Potassium dichromate 0.02 mol/l (0.12 N) in aqueous solution
Catalog # ROTHP720.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

About this item

 :
 :294.18 g/mol
Storage Temperature:Ambient
EINECS:231-906-6
UN:3287
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367