Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
 
n-Hexane ≥95%, HiPerSolv CHROMANORM® for HPLC
  83991.320
 :  
undefined
 
 
 
 
n-Hexane ≥95%, HiPerSolv CHROMANORM® for HPLC
  83991.320
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay (on anhydrous substance):
    Min. 95.0 %
  • Acidity:
    Max. 0.0005 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.01 %
  • Transmittance (200 nm):
    Min. 50 %
  • Transmittance (210 nm):
    Min. 70 %
  • Transmittance (220 nm):
    Min. 90 %
  • Transmittance (230 nm):
    Min. 98 %
  • Transmittance (240 nm):
    Min. 99 %

 

 

 :110-54-3
 :
 :H₃C(CH₂)₄CH₃
 :MFCD00009520
 :86.18 g/mol
 :13,04712
 :69 °C (1013 hPa)
 :–94,3 °C
 :0.659 g/cm³ (20 °C)
 :Ambient
 :203-777-6
 :1208
 :3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
 :–22 °C
 :01-2119480412-44
 :GHS02
 :GHS08
 :GHS06
 :GHS09
 :92112-69-1