Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.714 mol/l (0.714 N) in aqueous solution, AVS TITRINORM volumetric solution
Corrosive
Sodium hydroxide 0.714 mol/l (0.714 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30028.400
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium hydroxide 0.714 mol/l (0.714 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30028.400
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Titer (20°C;actual value 0,2 % accuracy):
    0,7125 - 0,7155 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :40 g/mol
Density:1.030 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548