Sodium metaarsenite 0.1 M (0.2 N) in aqueous solution
Catalog # 309744.1000
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Sodium dioxoarsenate
:
:129.91 g/mol
Storage Temperature:Ambient
EINECS:232-070-5
UN:1686
ADR:6.1,II
:GHS06
:GHS09
CAS Number:7784-46-5
Formula:NaAsO₂
MDL Number:MFCD00003472