Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sodium deoxycholate

Glass bottle for solids, 100 g
N° de catalogue MOLE13082357-100G
Irritant
Marketsource
Fournisseur:   Molekula
Sodium deoxycholate
N° de catalogue MOLE13082357-100G
Fournisseur:  Molekula
Marketsource
Numéro CAS:  302-95-4
Sodium deoxycholate

Glass bottle for solids, 100 g

Numéro CAS:  302-95-4
Prix
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  • CAS Number:302-95-4
  • Formula:C₂₄H₃₉NaO₄
  • MDL Number:MFCD00064139
  • Molecular Weight:414.56 g/mol
  • Melting Pt:357…365 °C
  • Storage Temperature:Ambient
  • EINECS:206-132-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications