Thymine
Glass bottle for solids, 25 g
MOLE31681796-25G
Marketsource
: Molekula
: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
- :65-71-4
- :C₅H₆N₂O₂
- :MFCD00006026
- :126.11 g/mol
- :12,09539
- :389 °C (1013 hPa)
- :218 °C
- :1.454 g/cm³ (25 °C)
- :200-616-1
- Pack type:Glass bottle for solids
- Pk:25 g