Order Entry
Belgium
ContactUsLinkComponent
Methyl hydrogen adipate
Methyl hydrogen adipate
Catalog # MOLE41497308-25G
Supplier:  Molekula
CAS Number:  
Methyl hydrogen adipate
Catalog # MOLE41497308-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :160.17 g/mol
Boiling Pt:292 °C (1013 hPa)
Density:1.1236 g/cm³ (20 °C)
EINECS:211-019-0
CAS Number:627-91-8
Formula:C₇H₁₂O₄
MDL Number:MFCD00004418