Order Entry
Belgium
ContactUsLinkComponent
Sodium nitroprusside dihydrate
Toxic
Sodium nitroprusside dihydrate
Catalog # MOLEM53304842
Supplier:  Molekula
CAS Number:  
Toxic
Sodium nitroprusside dihydrate
Catalog # MOLEM53304842
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Merck Index:13,08722
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1588
ADR:6.1,III
 :297.95 g/mol
EINECS:238-373-9
 :GHS06
CAS Number:13755-38-9
Formula:Na₂[Fe(CN)₅NO]·2H₂O
MDL Number:MFCD00149192