Neutral red
Glass bottle for solids, 25 g
Catalog # MOLEM27400744
Marketsource
Supplier: Molekula
Aller à :
Product Details & Documents
Synonymes: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
- Numéro CAS:553-24-2
- Formule:C₁₅H₁₇ClN₄
- Numéro MDL:MFCD00012651
- Poids moléculaire:288.78 g/mol
- Index Merck:12,06571
- Point de fusion:290 °C
- Température de stockage:Ambient
- EINECS:209-035-8
Spécifications
Spécifications
- Pack type:Glass bottle for solids
- Pk:25 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



