Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-N-(trimethylsilyl)trifluoroacetamide for synthesis, Sigma-Aldrich®
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Molar Mass:
    199.25 g/mol
  • Boiling Point:
    130 - 132 °C (1013 hPa)
  • Flash Point:
    25 °C
  • Density:
    1.085 g/cm3

Specification Test Results

Related Information

Frequently Bought Together