


Formaldehyde 37% in aqueous solution stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ200
:Stabilised with methanol 10 %
EINECS:200-001-8
UN:2209
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Density:1.017 g/cm3 (20 °C)
- pH:0.5 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Frequently Bought Together