Order Entry
Belgium
ContactUsLinkComponent
Tetrakis(triphenylphosphine)palladium(0) for synthesis, Sigma-Aldrich®
Irritant
Tetrakis(triphenylphosphine)palladium(0) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Tetrakis(triphenylphosphine)palladium(0) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    103 - 107 °C
  • Molar Mass:
    1155.58 g/mol
  • Hill Formula:
    C72H60P4Pd
  • Chemical Formula:
    Pd(P(C6H5)3)4

Specification Test Results

Related Information

Frequently Bought Together