Order Entry
Belgium
ContactUsLinkComponent
Palladium(II) nitrate dihydrate for synthesis, Sigma-Aldrich®
Oxidizing
Toxic
Corrosive
Environment
Palladium(II) nitrate dihydrate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Oxidizing
Toxic
Corrosive
Environment
Palladium(II) nitrate dihydrate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (slow decomposition)
  • Molar Mass:
    266.44 g/mol
  • pH:
    1 (1 g/l, H2O, 20 °C)
  • Hill Formula:
    N2O6Pd * 2 H2O
  • Chemical Formula:
    Pd(NO3)2 * 2 H2O

Specification Test Results

Related Information

Frequently Bought Together