Allylthiourea ≥98.0% for synthesis, Sigma-Aldrich®
: Merck
:109-57-9
:
:H₂CCHCH₂NHC(S)NH₂
:MFCD00004940
:116.19 g/mol
:12,09500
:191 °C (1013 hPa)
:74 °C
:1.219 g/cm³ (20 °C)
:2811
:6.1,III
:M-Clarity™ quality level = MQ200
:203-683-5
:GHS06
:Ambient
- CAS:109-57-9
- Synonyms:Thiosinamine
- Hill Formula:C₄H₈N₂S
- Molar Mass:116.19 g/mol
- Melting Point:70 - 73 °C
- Bulk density:550 kg/m³
- Solubility:66 g/l (20 °C)
- LD 50 oral:LD50 Rat 200 mg/kg
- Assay (HPLC, area%):≥ 98.0 % (a/a)
- Identity (IR):passes test
Frequently Bought Together