Order Entry
Belgium
ContactUsLinkComponent
Phenol red, EMPROVE® ESSENTIAL, SAFC®
Phenol red, EMPROVE® ESSENTIAL, SAFC®
Catalog # 1.37038.0100
CAS Number:  
Phenol red, EMPROVE® ESSENTIAL, SAFC®
Catalog # 1.37038.0100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Merck Index:13,07329
Melting Pt:> 300 °C
 :354.38 g/mol
Storage Temperature:Ambient
EINECS:205-609-7
CAS Number:143-74-8
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552