

N,O-Bis(trimethylsilyl)trifluoroacetamide ≥98.0% (by GC) derivatisation reagent, for gas chromatography, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:25561-30-2
:
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269
:257.4 g/mol
Boiling Pt:145 °C (1013 hPa)
Melting Pt:–41 °C
Density:0.96 g/cm³ (20 °C)
Flash Pt:34 °C
UN:1993
ADR:3,
:M-Clarity™ quality level = MQ100
EINECS:247-103-9
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (GC, area%):≥ 98.0 % (a/a)
- Identity (IR):Passes test
Specification Test Results
Related Information
Frequently Bought Together