Order Entry
Belgium
ContactUsLinkComponent
 
Sodium carbonate 0.05 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09940.0001
Supplier:  Merck
CAS Number:  
undefined
Sodium carbonate 0.05 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09940.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.1 g/cm3 (20 °C)
  • pH:
    11.6 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :105.99 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:207-838-8
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494