Order Entry
Belgium
Orders LinkContactUsLinkComponent
Neutral red

Neutral red

1 kg
Catalog # 1.01369.1000
Supplier:   Merck
Neutral red
Catalog # 1.01369.1000
Supplier:  Merck
CAS Number:  553-24-2
Neutral red

1 kg

CAS Number:  553-24-2
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS Number:553-24-2
  • Formula:C₁₅H₁₇ClN₄
  • MDL Number:MFCD00012651
  • Molecular Weight:288.78 g/mol
  • Merck Index:12,06571
  • Melting Pt:290 °C
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:209-035-8

Documents

Specifications

Specifications

  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.