Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sodium acetate 20% in aqueous solution, extra pure

Supplier:   Bernd Kraft
Sodium acetate 20% in aqueous solution, extra pure
Supplier:  Bernd Kraft
Certificates
Sodium acetate 20% in aqueous solution, extra pure

3 Options Available Below

Product Details & Documents
Synonyms:  Acetic acid sodium salt
  • CAS Number:127-09-3
  • Formula:H₃CCOONa
  • MDL Number:MFCD00012459
  • Molecular Weight:82.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:204-823-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    0,5 L
    Log in for availability
    Log in for availability
    1 * 0,5 L
    1 * 0,5 L

  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 65,40 €
    65,40 €
    1 * 1 L
       65,40 €

  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L