EDTA disodium salt 0.01 mol/l (0.02 N) in aqueous solution
Supplier: FF CHEMICALS
Certificates
About this item
CAS Number:139-33-3
:
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
:336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
Frequently Bought Together