1-(4-Fluorobenzyl)homopiperazin
Catalog # MAYBAW00512.0001
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:208.28 g/mol
CAS Number:76141-89-4
Formula:C₁₂H₁₇FN₂
MDL Number:MFCD00816588