Human thrombin (from plasma)
Glass bottle, 1 mL
Catalog # 605204-1
Supplier: Merck Millipore (Calbiochem)
Jump to:
Product Details & Documents
Native thrombin from human plasma, bound to agarose beads.
- Immobilised
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Cat. No.:605204-1
- Species:Human
- Source:Plasma
- Enzyme name:Thrombin
- CAS No.:9002-04-4



