

5-Fluorouracil, Millipore®
Catalog # 343922-1
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:130.08 g/mol
Merck Index:13,04208
Boiling Pt:347 °C (1013 hPa)
Melting Pt:282…283 °C
Storage Temperature:Ambient
EINECS:200-085-6
UN:2811
ADR:6.1,III
:GHS06
:GHS08
CAS Number:51-21-8
Formula:C₄H₃FN₂O₂
MDL Number:MFCD00006018