Order Entry
Belgium
Orders LinkContactUsLinkComponent
Propidium Iodide

Propidium Iodide

Glass bottle for solids, 100 mg
Catalog # 40016.
Irritant
Supplier:   Biotium
Propidium Iodide
Catalog # 40016.
Supplier:  Biotium
CAS Number:  25535-16-4
Propidium Iodide

Glass bottle for solids, 100 mg

CAS Number:  25535-16-4
Price
149,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Propidium iodide (PI) is a membrane-impermeant nucleic acid intercalator. The dye is commonly used to selectively stain dead cells in a cell population and also used as a counterstain in multicolor fluorescent imaging.

  • λEx/λEm (DNA-bound) = 535/617 nm
  • λEx/λEm (no DNA) = 493/636 nm
  • Orange red solid soluble in H2O or DMSO
  • Store at 4°C and protect from light, especially in solution

Propidium iodide (PI) is a cell impermeable nucleic acid intercalating dye. Because PI is excluded from viable cells, it can be used to distinguish necrotic or late apoptotic cells with damaged plasma membranes from viable cells or early apoptotic cells with intact cell membranes. After entering cells and binding to DNA and RNA, the fluorescence of PI is enhanced 20- to 30-fold. PI can be used in flow cytometry and fluorescence microscopy. PI is also utilized as a counterstain in multicolor fluorescent imaging, but nuclear-specific staining requires digestion of cellular RNA.

Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
  • EINECS:247-081-0

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.