7-Nitro-1H-indazole
Catalog # 00240.
Supplier: Biotium
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:163.14 g/mol
Melting Pt:190 °C
Density:1,56 g/cm³ (293 °C)
EINECS:220-934-4
CAS Number:2942-42-9
Formula:C₇H₅N₃O₂
MDL Number:MFCD00022789