Order Entry
Belgium
ContactUsLinkComponent
EDTA disodium salt concentrate for dilution to 0.01 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
EDTA disodium salt concentrate for dilution to 0.01 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4870.
Supplier:  Avantor
CAS Number:  
EDTA disodium salt concentrate for dilution to 0.01 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4870.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    60 mL
  • Seq:
    2385

Specifications

Specification Test Results

  • Titer (mol/l):
    0.0099-0.0101

Specification Test Results

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672