ContactUsLinkComponent
EDTA disodium salt concentrate for dilution to 0.1 M in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
  4653.
 :  Avantor
 :  
undefined
EDTA disodium salt concentrate for dilution to 0.1 M in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
  4653.
 :  Avantor
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • For Laboratory,Research,or Manufacturing Use:
  • Molarity (by titrimetry):
    Passes Test
  • (Makes 0.1M solution after dilution to 1000 mL):
 :
 :336.21 g/mol
 :12,03556
 :Ambient
 :205-358-3
 :139-33-3
 :C₁₀H₁₄N₂Na₂O₈
 :MFCD00070672