

2-Propanol USP, J.T.Baker®
: Avantor
:67-63-0
:
:(CH₃)₂CHOH
:MFCD00011674
:60.1 g/mol
:13,05228
:82 °C (1013 hPa)
:–89 °C
:0.786 g/cm³ (20 °C)
:12 °C
:Ambient
:200-661-7
:1219
:3,II
:01-2119457558-25
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Gravity at 25°/25°C:0.783 - 0.787
- USP - Refractive Index, n²ºD:1.376 - 1.378
- USP - Acidity:Passes Test
- USP - Nonvolatile Residue:≤0.005 %
- USP - Assay:≥99.0 %
- USP - Water (H₂O):≤0.5 %
- USP - Volatile Impurities - Ethyl ether:≤0.1 %
- USP - Volatile Impurities - Acetone:≤0.1 %
- USP - Volatile Impurities - Diisopropyl ether:≤0.1 %
- USP - Volatile Impurities - 1-Propanol:≤0.1 %
- USP - Volatile Impurities - 2-Butanol:≤0.1 %
- USP - Volatile Impurities - Total:≤1.0 %
- USP - Benzene:≤2 ppm
Frequently Bought Together