Order Entry
Belgium
ContactUsLinkComponent
Methanol ≥99.9% (by GC), CMOS for microelectronic, J.T.Baker®
Flammable
Toxic
Health
Methanol ≥99.9% (by GC), CMOS for microelectronic, J.T.Baker®
Catalog # 9073-05
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol ≥99.9% (by GC), CMOS for microelectronic, J.T.Baker®
Catalog # 9073-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, pressure bottom
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Microelectronic Use:
    = 99.9 %
  • Assay (CH₃OH) (by GC):
    ≥99.9 %
  • Color (APHA):
    ≤10
  • Acidity (µeq/g):
    ≤0.3
  • Alkalinity (µeq/g):
    ≤0.1
  • Heavy Metals (as Pb):
    ≤100.0 ppb
  • Residue after Evaporation:
    ≤5 ppm
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %
  • Solubility in H₂O:
    Passes Test
  • Chloride (Cl):
    ≤0.2 ppm
  • Phosphate (PO₄):
    ≤0.3 ppm
  • Sulfate (SO₄):
    ≤0.5 ppm
  • Trace Impurities - Aluminum (Al):
    ≤50.0 ppb
  • Trace Impurities - Arsenic (As):
    ≤5.0 ppb
  • Trace Impurities - Antimony (Sb):
    ≤5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤20.0 ppb
  • Trace Impurities - Boron (B):
    ≤10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤20.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤50.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤20.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤20 ppb
  • Trace Impurities - Copper (Cu):
    ≤10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤50 ppb
  • Trace Impurities - Germanium (Ge):
    ≤50.0 ppb
  • Trace Impurities - Gold (Au):
    ≤20 ppb
  • Trace Impurities - Iron (Fe):
    ≤50.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤50 ppb
  • Trace Impurities - Lithium (Li):
    ≤50.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤50.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤10.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤300.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤50 ppb
  • Trace Impurities - Silicon (Si):
    ≤50 ppb
  • Trace Impurities - Silver (Ag):
    ≤20.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤50.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤50 ppb
  • Trace Impurities - Titanium (Ti):
    ≤20 ppb
  • Trace Impurities - Vanadium (V):
    ≤300.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤50 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤300.0 ppb
  • Particle Count - 0.5 µm and greater:
    ≤100 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤8 par/ml

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595