Order Entry
Belgium
ContactUsLinkComponent
2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®
Flammable
Irritant
Health
2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Health
2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Specific Gravity at 25°/25°C:
    0.783 - 0.787
  • USP - Refractive Index, n²ºD:
    1.376 - 1.378
  • USP - Acidity:
    Passes Test
  • USP - Nonvolatile Residue:
    ≤0.005 %
  • USP - Assay:
    ≥99.0 %
  • USP - Water (H₂O):
    ≤0.5 %
  • USP - Volatile Impurities - Ethyl ether:
    ≤0.1 %
  • USP - Volatile Impurities - Acetone:
    ≤0.1 %
  • USP - Volatile Impurities - Diisopropyl ether:
    ≤0.1 %
  • USP - Volatile Impurities - 1-Propanol:
    ≤0.1 %
  • USP - Volatile Impurities - 2-Butanol:
    ≤0.1 %
  • USP - Volatile Impurities - Total:
    ≤1.0 %
  • USP - Benzene:
    ≤2 ppm
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • Appearance:
    Passes Test
  • EP/BP - Acidity or Alkalinity:
    Passes Test
  • EP/BP - Benzene (C₆H₆):
    ≤2 ppm
  • EP/BP - Any peak other than 2-butanol:
    ≤0.3 %
  • EP/BP - Peroxide:
    Passes Test
  • EP/BP - Nonvolatile Matter:
    ≤20 ppm
  • EP/BP - Water (H₂O):
    ≤0.5 %
  • EP/BP - UV Absorbance - 310 nm:
    ≤0.01
  • EP/BP - UV Absorbance - 290 nm:
    ≤0.02
  • EP/BP - UV Absorbance - 270 nm:
    ≤0.03
  • EP/BP - UV Absorbance - 250 nm:
    ≤0.10
  • EP/BP - UV Absorbance - 230 nm:
    ≤0.30
  • EP/BP - Spectrum Appearance:
    Passes Test
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Specific Gravity at 20°/20°C:
    0.785 - 0.788
  • JP - Clarity of Solution:
    Passes Test
  • JP - Acidity:
    Passes Test
  • JP - Residue on Evaporation, mg/20mL:
    ≤1.0
  • JP - Water (H₂O):
    ≤0.75 %
  • JP - Distilling Range(Lower):
    81 - 83 °C
  • JP - Distilling Range (Upper):
    81 - 83 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together