Order Entry
Belgium
ContactUsLinkComponent
Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis), granular NF, FCC, J.T.Baker®
Irritant
Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis), granular NF, FCC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis), granular NF, FCC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    Passes Test
  • Food GMP Manufactured Product:
    Passes Test
  • Meets F.C.C. Requirements:
    ≤0.5%
  • Meets N.F. Requirements:
    99.5 - 100.5%
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Food Chemical:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    ≤0.5%
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Water (H₂O):
    ≤0.5 %
  • NF - Assay (Na₂CO₃) (anhydrous basis):
    99.5 - 100.5 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Loss on Drying:
    ≤0.5 %
  • FCC - Assay (Na₂CO₃) (anhydrous basis):
    99.5 - 100.5 %
  • FCC - Lead (Pb):
    ≤4 mg/kg

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together