Chenodeoxycholic acid
Catalog # A1690.0005
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:392.58 g/mol
Merck Index:12,02096
Melting Pt:164.85…165.85 °C
Density:1.162 g/cm³ (20 °C)
EINECS:207-481-8
CAS Number:474-25-9
Formula:C₂₄H₄₀O₄
MDL Number:MFCD00064142