Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium deoxycholate

Sodium deoxycholate

Irritant
Supplier:   PanReac AppliChem
Sodium deoxycholate
Supplier:  PanReac AppliChem
Certificates
Sodium deoxycholate

2 Options Available Below

Product Details & Documents
Synonyms:  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  • CAS Number:302-95-4
  • Formula:C₂₄H₃₉NaO₄
  • MDL Number:MFCD00064139
  • Molecular Weight:414.56 g/mol
  • Melting Pt:357…365 °C
  • Storage Temperature:Ambient
  • EINECS:206-132-7

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 144,00 €
    144,00 €
    1 * 100 g
       144,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1 230,00 €
    1 230,00 €
    1 * 1 kg
       1 230,00 €