Order Entry
Belgium
ContactUsLinkComponent
VWR® Pepsin
VWR® Pepsin
Catalog # M142-250G
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Pepsin
Catalog # M142-250G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • CAS No.:
    9001-75-6
  • Storage conditions:
    Refrigerator
  • Enzyme name:
    Pepsin
  • Enzyme activity:
    1:3.000
  • Grade:
    Proteomics grade

Specifications

About this item