Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(-)-Ribose ≥98%

D-(-)-Ribose ≥98%

D-(-)-Ribose ≥98%
Certificates
D-(-)-Ribose ≥98%
D-(-)-Ribose ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 50-69-1 Molecular Formula: C5H10O5 Formula Weight: 150.13 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 82-86° Beilstein Reference: 1723081 Merck Reference: 14,8204 Optical Rotation: -20° (c=10 in water, 28h) MDL No.: MFCD00135453

  • CAS Number:50-69-1
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00135453
  • Molecular Weight:150.13 g/mol
  • Merck Index:14,8204
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:82…86 °C
  • Density:0.8 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-059-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 33,00 €
    33,00 €
    1 * 10 g
       33,00 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 80,10 €
    80,10 €
    1 * 50 g
       80,10 €

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 362,00 €
    362,00 €
    1 * 250 g
       362,00 €