Order Entry
Belgium
Orders LinkContactUsLinkComponent
Thymine 97%
Thymine 97%
Certificates
Thymine 97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 65-71-4 Molecular Formula: C5H6N2O2 Formula Weight: 126.12 Melting Point: 316-317° Beilstein Reference: 607626 Merck Reference: 14,9398 MDL No.: MFCD00006026

Synonyms:  5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
  • CAS Number:65-71-4
  • Formula:C₅H₆N₂O₂
  • MDL Number:MFCD00006026
  • Molecular Weight:126.11 g/mol
  • Merck Index:14,9398
  • Boiling Pt:389 °C (1013 hPa)
  • Melting Pt:316…317 °C
  • Density:1.454 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-616-1
Product Family Options

Product Information