Sodium hydroxide ≥99.99% (metals basis), beads/pellets
About this item
Purity: 99.99% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Density: 2.13 Sensitivity: Air Sensitive and Hygroscopic Appearance: White to off-white Odour: Odourless pH: 14 Melting Point: 316° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,8627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:316 °C
- Density:2.13
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
3 Options Available Below
Product Details & Documents
Purity: 99.99% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Density: 2.13 Sensitivity: Air Sensitive and Hygroscopic Appearance: White to off-white Odour: Odourless pH: 14 Melting Point: 316° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,8627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:316 °C
- Density:2.13
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Documents
Product Family Options
Product Information
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.



