

Adipoyl dichloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-50-2
:
Formula:C₆H₈Cl₂O₂
MDL Number:MFCD00000759
:183.03 g/mol
Boiling Pt:105…107 °C (2 mmHg)
Density:1.25 g/cm³
Flash Pt:72 °C
EINECS:203-876-4
UN:2922
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear brown Liquid
- Infrared spectrum:Conforms
- Titration RCOCl:≥97.5 %
- Titration RCOOH:≤2 % (as Adipic acid)
Specification Test Results
Frequently Bought Together