Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Thymine 99%

Irritant
Thymine 99%
Certificates
Thymine 99%

4 Options Available Below

Product Details & Documents
Synonyms:  5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
  • CAS Number:65-71-4
  • Formula:C₅H₆N₂O₂
  • MDL Number:MFCD00006026
  • Molecular Weight:126.11 g/mol
  • Merck Index:12,09539
  • Boiling Pt:389 °C (1013 hPa)
  • Melting Pt:316…317 °C
  • Density:1,454 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-616-1

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥98.5 %
  • Chloride (Cl):
    ≤0.005 %
  • Sulfate (SO4):
    ≤0.01 %
  • Calcium (Ca):
    ≤0.002 %
  • Iron (Fe):
    ≤0.002 %
  • Sodium (Na):
    ≤0.005 %
  • Insoluble matter:
    ≤0.005 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 39,90 €
    39,90 €
    1 * 5 g
       39,90 €

  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 125,00 €
    125,00 €
    1 * 25 g
       125,00 €

  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 368,00 €
    368,00 €
    1 * 100 g
       368,00 €

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 320,00 €
    1 320,00 €
    1 * 500 g
       1 320,00 €