Orders LinkContactUsLinkComponent
Micro Curettes
Marketsource
Supplier:   FINE SCIENCE TOOLS
Micro Curettes
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
4 Options Available Below
Product Details & Documents
Stainless steel. A curette tip consists of a scoop with sharp edges and is used for scraping or cleaning. It also can be used for separating and transporting, inoculating plates or transferring small amounts of chemicals.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • FSCI10083-20
    Micro curette, tip diameter 2,0 mm, serrated
    Micro curette, tip diameter 2,0 mm, serrated
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 114,00 €
    114,00 €
    1 * 1 Pce
       114,00 €
    Specifications:
    Marketsource
    Marketsource
  • FSCI10082-15
    Micro curette, tip diameter 1,5 mm
    Micro curette, tip diameter 1,5 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 83,00 €
    83,00 €
    1 * 1 Pce
       83,00 €
    Specifications:
    Marketsource
    Marketsource
  • FSCI10081-10
    Micro curette, tip diameter 1,0 mm
    Micro curette, tip diameter 1,0 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 83,00 €
    83,00 €
    1 * 1 Pce
       83,00 €
    Specifications:
    Marketsource
    Marketsource
  • FSCI10080-05
    Micro curette, tip diameter 0,5 mm
    Micro curette, tip diameter 0,5 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 82,50 €
    82,50 €
    1 * 1 Pce
       82,50 €
    Specifications:
    Marketsource
    Marketsource