Order Entry
Belgium
ContactUsLinkComponent
Tetrakis(triphenylphosphine)palladium(0) ≥99% (99.9+%-Pd)
Tetrakis(triphenylphosphine)palladium(0) ≥99% (99.9+%-Pd)
Catalog # STRE46-2150-5G
CAS Number:  
Tetrakis(triphenylphosphine)palladium(0) ≥99% (99.9+%-Pd)
Catalog # STRE46-2150-5G
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :1155.58 g/mol
Melting Pt:103…107 °C
Density:1 g/cm³ (20 °C)
EINECS:238-086-9
CAS Number:14221-01-3
Formula:C₇₂H₆₀P₄Pd
MDL Number:MFCD00010012