Order Entry
Belgium
Orders LinkContactUsLinkComponent
Propidium Iodide

Propidium Iodide

Glass bottle, 1 mL
Catalog # ABCAAB14083-1
Irritant
Marketsource
Supplier:   Abcam
Propidium Iodide
Catalog # ABCAAB14083-1
Supplier:  Abcam
Marketsource
CAS Number:  25535-16-4
Propidium Iodide

Glass bottle, 1 mL

CAS Number:  25535-16-4
Price
175,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
  • EINECS:247-081-0
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications