Orders LinkContactUsLinkComponent
Skin, block model
Skin, block model
Skin structure
Catalog # 763-0123
Marketsource
Supplier:   3B Scientific
Skin, block model
Catalog # 763-0123
Supplier:  3B Scientific
Marketsource
Skin, block model
Skin, block model
Skin structure
Price
288,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
70× enlargement in the model of skin with hair from the head, skin with hair from the armpit and skin from a finger. Sweat glands, sensory bodies, etc. are inlaid in the three sections. Dimensions: 30×36×32 cm.
Specifications
  • Description:
    Skin structure